2-Nitro-4-(trifluoromethyl)phenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Hydroxy-3-nitrobenzotrifluoride
L10604-03
L10604-06
CAAAAL10604-03EA
80.26
CAD
CAAAAL10604-03
CAAAAL10604-06
2-Nitro-4-(trifluoromethyl)phenol 98%
2-Nitro-4-(trifluoromethyl)phenol
Purity: 98%
CAS No.: 400-99-7
Molecular Formula: C7H4F3NO3
Formula Weight: 207.11
Flash Point: 95°(203°F)
Refractive Index: 1.5040
Boiling Point: 92-94°/12mm
Beilstein Reference: 2215510
MDL No.: MFCD00009791
Formula:
C₇H₄F₃NO₃ MW: 207.11 Boiling Pt: 92…94 °C (12 mmHg) Density: 1.473 Flash Pt: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009791 CAS Number: 400-99-7 EINECS: 206-927-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...