Propagyl bromide ≥80% (w/w) in toluene stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromopropyne

Stabilization: Stabiised with magnesium oxide (MgO)

L10595-14 L10595-22
CAAAAL10595-14EA 70.47 CAD
CAAAAL10595-14 CAAAAL10595-22
Propagyl bromide ≥80% (w/w) in toluene stabilized
Propagyl bromide

Purity: 97% CAS No.: 106-96-7 Molecular Formula: C3H3Br Formula Weight: 118.97 Storage Temperature: 2°C to 8°C Flash Point: 3°(37°F) Refractive Index: 1.4940 Melting Point: -61° Boiling Point: 88-90° Beilstein Reference: 605309 UN No.: 2924 MDL No.: MFCD00000241

Formula: C₃H₃Br
MW: 118.96 g/mol
Boiling Pt: 88…90 °C
Melting Pt: –61 °C
Density: 1.335
Flash Pt: 3 °C (37 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000241
CAS Number: 106-96-7
EINECS: 203-447-1
UN: 3286
ADR: 3 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR