2-(Trifluoromethyl)phenyl isothiocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10586-03
L10586-06
CAAAAL10586-03EA
46.45
CAD
CAAAAL10586-03
CAAAAL10586-06
2-(Trifluoromethyl)phenyl isothiocyanate 98%
2-(Trifluoromethyl)phenyl isothiocyanate
Purity: 98%
CAS No.: 1743-86-8
Molecular Formula: C8H4F3NS
Formula Weight: 203.19
Flash Point: 105°(221°F)
Refractive Index: 1.5660
Odor: Pungent
Boiling Point: 216°
Beilstein Reference: 2693552
UN No.: 2922
MDL No.: MFCD00039644
Formula:
C₈H₄F₃NS Boiling Pt: 216 °C Density: 1.346 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039644 CAS Number: 1743-86-8 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...