1-Bromo-3,3,3-trifluoro-2-propanol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Bromo-1,1,1-trifluoro-2-propanol
L10566-03
L10566-06
CAAAAL10566-03EA
68.89
CAD
CAAAAL10566-03
CAAAAL10566-06
1-Bromo-3,3,3-trifluoro-2-propanol 96%
1-Bromo-3,3,3-trifluoro-2-propanol
Purity: 96%
CAS No.: 431-34-5
Molecular Formula: C3H4BrF3O
Formula Weight: 192.97
Flash Point: 43°(109°F)
Refractive Index: 1.3990
Boiling Point: 124-126°
Beilstein Reference: 1737803
UN No.: 1987
MDL No.: MFCD00040945
Formula:
BrH₂CCH(OH)CF₃ Boiling Pt: 124…126 °C Density: 1.861 Flash Pt: 43 °C(109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040945 CAS Number: 431-34-5 UN: 1987 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...