2-Chlorobenzyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Chloro-2-(isothiocyanatomethyl)benzene
L10546-03
L10546-06
CAAAAL10546-03EA
39.18
CAD
CAAAAL10546-03
CAAAAL10546-06
2-Chlorobenzyl isothiocyanate 97%
2-Chlorobenzyl isothiocyanate
Purity: 97%
CAS No.: 18967-44-7
Molecular Formula: C8H6ClNS
Formula Weight: 183.66
Storage Temperature: 2°C to 8°C
Refractive Index: 1.6140
Odor: Irritating
Boiling Point: 137-138°/6mm
Beilstein Reference: 2936688
UN No.: 2922
MDL No.: MFCD00041110
Formula:
C₈H₆ClNS MW: 183.66 g/mol Boiling Pt: 137…138 °C (6 mmHg) Density: 1.149 Storage Temperature: Refrigerator |
MDL Number:
MFCD00041110 CAS Number: 18967-44-7 UN: 2922 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...