trans-3-Hexen-1-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
trans-Hex-3-en-1-ol
L10541-06
L10541-14
CAAAAL10541-06EA
84.21
CAD
CAAAAL10541-06
CAAAAL10541-14
trans-3-Hexen-1-ol 97%
trans-3-Hexen-1-ol
Purity: 97%
CAS No.: 928-97-2
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 58°(136°F)
Refractive Index: 1.4390
Odor: Pleasant
Boiling Point: 60-61°/12mm
Beilstein Reference: 1719713
Merck Reference: 14,4700
UN No.: 1987
MDL No.: MFCD00002960
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 60…61 °C/12 mm Melting Pt: –60 °C Density: 0.845 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002960 CAS Number: 928-97-2 EINECS: 213-193-3 UN: 1987 ADR: 3,III Merck Index: 14,4700 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...