1-(2-Hydroxyphenyl)-2-thiourea 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-(2-Hydroxyphenyl)thiourea, 2-Hydroxyphenylthiourea
L10519-03
L10519-06
CAAAAL10519-03EA
33.18
CAD
CAAAAL10519-03
CAAAAL10519-06
1-(2-Hydroxyphenyl)-2-thiourea 97%
1-(2-Hydroxyphenyl)-2-thiourea
Purity: 97%
CAS No.: 1520-26-9
Molecular Formula: C7H8N2OS
Formula Weight: 168.22
Odor: Odorless
Melting Point: ca 158° dec.
Beilstein Reference: 1102479
MDL No.: MFCD00041159
Formula:
(C₆H₄(OH))NC(S)NH₂ MW: 168.22 g/mol Boiling Pt: 323 Melting Pt: 162 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00041159 CAS Number: 1520-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...