1,3-Cyclooctadiene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cycloocta-1,3-diene
L10516-06
L10516-14
CAAAAL10516-06EA
53.56
CAD
CAAAAL10516-06
CAAAAL10516-14
1,3-Cyclooctadiene 97%
1,3-Cyclooctadiene
Purity: 97%
CAS No.: 1700-10-3
Molecular Formula: C8H12
Formula Weight: 108.18
Storage Temperature: 2°C to 8°C
Flash Point: 24°(75°F)
Refractive Index: 1.4930
Odor: Characteristic
Melting Point: -53°
Boiling Point: 142-144°
Beilstein Reference: 1901033
UN No.: 2520
MDL No.: MFCD00001751
Formula:
C₈H₁₂ MW: 108.18 g/mol Boiling Pt: 142…144 °C Melting Pt: –53 °C Density: 0.869 Flash Pt: 24 °C (75 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001751 CAS Number: 1700-10-3 EINECS: 216-929-1 UN: 2520 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...