4-Isopropylbenzenesulfonyl chloride 96%

Supplier: Thermo Scientific Chemicals
Danger

L10428-06 L10428-14
CAAAAL10428-06EA 67.31 CAD
CAAAAL10428-06 CAAAAL10428-14
4-Isopropylbenzenesulfonyl chloride 96%
4-Isopropylbenzenesulphonyl chloride

Purity: 96% CAS No.: 54997-90-9 Molecular Formula: C9H11ClO2S Formula Weight: 218.70 Flash Point: 110°(230°F) Refractive Index: 1.5390 Boiling Point: 142°/5mm Beilstein Reference: 1103862 UN No.: 3265 MDL No.: MFCD00041510

Formula: C₉H₁₁ClO₂S
Boiling Pt: 142 °C/5 mm
Density: 1.220
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041510
CAS Number: 54997-90-9
UN: 3265
ADR: "8",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR