(S)-(+)-2-Hexanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(+)-Butyl methyl carbinol, (S)-(+)-Methyl butyl carbinol, (S)-(+)-2-Hydroxy hexane

L10401-03 L10401-MD
CAAAAL10401-03EA 225.94 CAD
CAAAAL10401-03 CAAAAL10401-MD
(S)-(+)-2-Hexanol 98%
(S)-(+)-2-Hexanol

Purity: 98% CAS No.: 52019-78-0 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 45°(113°F) Refractive Index: 1.4140 Boiling Point: 137-138° Beilstein Reference: 1718997 Optical Rotation: +10.5° (neat) UN No.: 2282 MDL No.: MFCD00065955

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 137
Density: 0.818
Flash Pt: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00065955
CAS Number: 52019-78-0
UN: 2282
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR