1,3-Cyclopentanedione 98%
Supplier: Thermo Scientific Chemicals
L10336-03
L10336-06
CAAAAL10336-03EA
79
CAD
CAAAAL10336-03
CAAAAL10336-06
1,3-Cyclopentanedione 98%
1,3-Cyclopentanedione
Purity: 98%
CAS No.: 3859-41-4
Molecular Formula: C5H6O2
Formula Weight: 98.10
Melting Point: 151-153°
Beilstein Reference: 1362728
Formula:
C₅H₆O₂ Melting Pt: 151…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001405 CAS Number: 3859-41-4 EINECS: 223-372-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...