trans-3-Hexene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(E)-3-Hexene, 3-Hexene
L10311-03
L10311-06
CAAAAL10311-03EA
63.83
CAD
CAAAAL10311-03
CAAAAL10311-06
trans-3-Hexene 98%
trans-3-Hexene
Purity: 98%
CAS No.: 13269-52-8
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -25°(-13°F)
Refractive Index: 1.3940
Odor: Mild
Melting Point: -113°
Boiling Point: 66-67°
Beilstein Reference: 1718859
UN No.: 3295
MDL No.: MFCD00009386
Formula:
C₆H₁₂ Boiling Pt: 66…67 °C Melting Pt: –113 °C Density: 0.677 Flash Pt: –25 °C (–13 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009386 CAS Number: 13269-52-8 EINECS: 236-261-4 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...