(R)-(+)-1-Phenylethyl isocyanate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(+)-α-Phenethyl isocyanate, (R)-(+)-1-Phenethyl isocyanate, (R)-(+)-1-phenylethylisocyanate, (R)-(+)-α-Methylbenzyl isocyanate
L10305-03
L10305-06
CAAAAL10305-03EA
81.05
CAD
CAAAAL10305-03
CAAAAL10305-06
(R)-(+)-1-Phenylethyl isocyanate 99%
(R)-(+)-1-Phenylethyl isocyanate
Purity: 99%
CAS No.: 33375-06-3
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.5124
Boiling Point: 55-56°/2.5mm
Beilstein Reference: 3196930
Optical Rotation: +10.5° (neat)
UN No.: 2206
MDL No.: MFCD00063015
Formula:
C₉H₉NO Boiling Pt: 55…56 °C/2.5 mm Density: 1.045 Flash Pt: 65 °C (149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD06739387 CAS Number: 33375-06-3 EINECS: 251-485-2 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...