5-Methoxyanthranilic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10295-03
L10295-06
CAAAAL10295-03EA
141.57
CAD
CAAAAL10295-03
CAAAAL10295-06
5-Methoxyanthranilic acid 98+%
5-Methoxyanthranilic acid
Purity: 98+%
CAS No.: 6705-03-9
Molecular Formula: C8H9NO3
Formula Weight: 167.16
Melting Point: 150-152°
Beilstein Reference: 777690
Formula:
C₈H₉NO₃ Melting Pt: 150…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016509 CAS Number: 6705-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...