Tetradecyltrimethylammonium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10294-22
L10294-36
CAAAAL10294-22EA
86.9
CAD
CAAAAL10294-22
CAAAAL10294-36
Tetradecyltrimethylammonium bromide 98%
Tetradecyltrimethylammonium bromide
Phase transfer catalyst
Purity: 98%
CAS No.: 1119-97-7
Molecular Formula: C17H38BrN
Formula Weight: 336.40
Odor: Odorless
Melting Point: 245-250°
Beilstein Reference: 3633227
Merck Reference: 14,6336
UN No.: 1759
MDL No.: MFCD00011770
Formula:
C₁₇H₃₈BrN Melting Pt: 245…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011770 CAS Number: 1119-97-7 EINECS: 214-291-9 UN: 1759 ADR: 8,III Merck Index: 14,6336 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...