(±)-2-Mercaptopropionic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Mercaptopropionic acid

L10257-09 L10257-22 L10257-36
CAAAAL10257-09EA 48.98 CAD
CAAAAL10257-09 CAAAAL10257-22 CAAAAL10257-36
(±)-2-Mercaptopropionic acid 97%
(±)-2-Mercaptopropionic acid

Purity: 97% CAS No.: 79-42-5 Molecular Formula: C3H6O2S Formula Weight: 106.14 Flash Point: 107°(225°F) Refractive Index: 1.4820 Odor: Unpleasant Melting Point: 10-14° Boiling Point: 217° Beilstein Reference: 506218 Merck Reference: 14,9340 UN No.: 2936 MDL No.: MFCD00004862

Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 217 °C
Melting Pt: 10…14 °C
Density: 1.196
Flash Pt: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004862
CAS Number: 79-42-5
EINECS: 201-206-5
UN: 2936
ADR: 6.1,II
Merck Index: 14,9340

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR