3-Hydroxy-p-anisidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxy-4-anisidine, 5-Amino-2-methoxyphenol, 3-Hydroxy-4-methoxyaniline, 4-Amino-2-hydroxyanisole

L10234-06 L10234-14
CAAAAL10234-06EA 111.07 CAD
CAAAAL10234-06 CAAAAL10234-14
3-Hydroxy-p-anisidine 98%
3-Hydroxy-p-anisidine

Purity: 98% CAS No.: 1687-53-2 Molecular Formula: C7H9NO2 Formula Weight: 139.16 Melting Point: 130-132° Beilstein Reference: 2717084

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Boiling Pt: 277 °C (1013 hPa)
Melting Pt: 130…132 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010222
CAS Number: 1687-53-2
EINECS: 216-873-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR