2,3-Dimethylphenyl isocyanate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Isocyanato-2,3-dimethylbenzene, 2,3-Xylyl isocyanate
L10225-03
L10225-06
CAAAAL10225-03EA
42.5
CAD
CAAAAL10225-03
CAAAAL10225-06
2,3-Dimethylphenyl isocyanate 99%
2,3-Dimethylphenyl isocyanate
Purity: 99%
CAS No.: 1591-99-7
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 92°(197°F)
Refractive Index: 1.5400
Odor: Pungent
Boiling Point: 104-105°/50mm
Beilstein Reference: 6313438
UN No.: 2206
MDL No.: MFCD00013851
Formula:
C₉H₉NO MW: 147.18 g/mol Boiling Pt: 104…105 °C (50 mmHg) Density: 1.059 Flash Pt: 92 °C (197 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013851 CAS Number: 1591-99-7 UN: 2206 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...