4,4-Dimethyl-2-pentyne 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10210-03
L10210-06
CAAAAL10210-03EA
96.54
CAD
CAAAAL10210-03
CAAAAL10210-06
4,4-Dimethyl-2-pentyne 97+%
4,4-Dimethyl-2-pentyne
Purity: 97+%
CAS No.: 999-78-0
Molecular Formula: C7H12
Formula Weight: 96.17
Flash Point: -10°(14°F)
Refractive Index: 1.4085
Odor: Petrol-like
Boiling Point: 82°
Beilstein Reference: 1733540
UN No.: 3295
MDL No.: MFCD00041613
Formula:
C₇H₁₂ Boiling Pt: 82 °C Melting Pt: –82.4 °C Density: 0.718 Flash Pt: –10 °C (14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041613 CAS Number: 999-78-0 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...