4-Cyanophenyl isothiocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10173-03
L10173-06
CAAAAL10173-03EA
48.35
CAD
CAAAAL10173-03
CAAAAL10173-06
4-Cyanophenyl isothiocyanate 98%
4-Cyanophenyl isothiocyanate
Purity: 98%
CAS No.: 2719-32-6
Molecular Formula: C8H4N2S
Formula Weight: 160.20
Odor: Irritating
Melting Point: 118-122°
Beilstein Reference: 2802621
UN No.: 2923
MDL No.: MFCD00041085
Formula:
C₈H₄N₂S Boiling Pt: 294 °C (1013 hPa) Melting Pt: 118…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041085 CAS Number: 2719-32-6 EINECS: 220-323-2 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...