(4-Nitrophenyl)-β-D-glucopyranoside 98+%

Supplier: Thermo Scientific Chemicals

L10154-03 L10154-06
CAAAAL10154-03EA 141.73 CAD
CAAAAL10154-03 CAAAAL10154-06
(4-Nitrophenyl)-β-D-glucopyranoside 98+%
(4-Nitrophenyl) β-D-glucopyranoside

Purity: 98+% CAS No.: 2492-87-7 Molecular Formula: C12H15NO8 Formula Weight: 301.25 Storage Temperature: 2°C to 8°C Melting Point: 166-167° Beilstein Reference: 92211 Optical Rotation: -103° (c=1 in water) MDL No.: MFCD00006593

Formula: C₁₂H₁₅NO₈
Melting Pt: 166…167 °C
Storage Temperature: Freezer
MDL Number: MFCD00006593
CAS Number: 2492-87-7
EINECS: 219-661-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR