4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L10127-03
L10127-06
CAAAAL10127-03EA
68.1
CAD
CAAAAL10127-03
CAAAAL10127-06
4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate 97%
4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate
Purity: 97%
CAS No.: 23163-86-2
Molecular Formula: C8H3ClF3NS
Formula Weight: 237.63
Flash Point:
Formula:
C₈H₃ClF₃NS Boiling Pt: 248…251 °C Melting Pt: 34…36 °C Density: 1.470 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041090 CAS Number: 23163-86-2 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...