4-Chloro-3-nitrophenol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10108-06
L10108-14
CAAAAL10108-06EA
64.94
CAD
CAAAAL10108-06
CAAAAL10108-14
4-Chloro-3-nitrophenol 99%
4-Chloro-3-nitrophenol
Purity: 99%
CAS No.: 610-78-6
Molecular Formula: C6H4ClNO3
Formula Weight: 173.56
Odor: Characteristic
Melting Point: 124-129°
Beilstein Reference: 2094546
MDL No.: MFCD00043546
Formula:
C₆H₄ClNO₃ Melting Pt: 124…129 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00043546 CAS Number: 610-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...