(S)-(+)-sec-Butylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-(+)-2-Aminobutane, (S)-(+)-2-Butylamine

L10069-03 L10069-06 L10069-MD
CAAAAL10069-03EA 257.54 CAD
CAAAAL10069-03 CAAAAL10069-06 CAAAAL10069-MD
(S)-(+)-sec-Butylamine 98%
(S)-(+)-sec-Butylamine

Purity: 98% CAS No.: 513-49-5 Molecular Formula: C4H11N Formula Weight: 73.14 Flash Point: -19°(-2°F) Refractive Index: 1.3930 Odor: Amine-like pH: 12.5 Boiling Point: 63° Beilstein Reference: 1718760 Merck Reference: 14,1544 Optical Rotation: +7.5° (neat) UN No.: 2733 MDL No.: MFCD00064417

Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 63 °C
Melting Pt: –104 °C
Density: 0.722
Flash Pt: –19 °C (–2 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064417
CAS Number: 513-49-5
EINECS: 208-164-7
UN: 3286
ADR: 3,II
Merck Index: 14,1544

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR