Diethyl disulfide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L10065-14
L10065-22
CAAAAL10065-14EA
89.59
CAD
CAAAAL10065-14
CAAAAL10065-22
Diethyl disulfide 99%
Diethyl disulphide
Purity: 99%
CAS No.: 110-81-6
Molecular Formula: C4H10S2
Formula Weight: 122.25
Flash Point: 40°(104°F)
Refractive Index: 1.5060
Odor: Stench
Melting Point: -102°
Boiling Point: 152-154°
Beilstein Reference: 1098273
UN No.: 1993
MDL No.: MFCD00009266
Formula:
C₄H₁₀S₂ Boiling Pt: 152…154 °C Melting Pt: –102 °C Density: 0.993 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009266 CAS Number: 110-81-6 EINECS: 203-805-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...