Sodium-3-nitrobenzoate, (max. 10% H₂O) 94%

Supplier: Thermo Scientific Chemicals
Danger

L10028-22 L10028-36
CAAAAL10028-22EA 62.57 CAD
CAAAAL10028-22 CAAAAL10028-36
Sodium-3-nitrobenzoate, (max. 10% H₂O) 94%
Sodium 3-nitrobenzoate

Purity: 94% CAS No.: 827-95-2 Molecular Formula: C7H4NNaO4 Formula Weight: 189.10 Melting Point: 300° Beilstein Reference: 4166479

Formula: C₇H₄NNaO₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150535
CAS Number: 827-95-2
EINECS: 212-578-3
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR