1-(3-Hydroxyphenyl)-2-thiourea 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09975-04
L09975-09
CAAAAL09975-04EA
119.45
CAD
CAAAAL09975-04
CAAAAL09975-09
1-(3-Hydroxyphenyl)-2-thiourea 97%
1-(3-Hydroxyphenyl)-2-thiourea
Purity: 97%
CAS No.: 3394-05-6
Molecular Formula: C7H8N2OS
Formula Weight: 168.22
Odor: Odorless
Melting Point: 175-180°
Beilstein Reference: 2090372
MDL No.: MFCD00022167
Formula:
(C₆H₄(OH))NC(S)NH₂ Melting Pt: 175…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022167 CAS Number: 3394-05-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...