2-Bromophenyl isothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

L09971-03 L09971-06
CAAAAL09971-03EA 33.65 CAD
CAAAAL09971-03 CAAAAL09971-06
2-Bromophenyl isothiocyanate 98%
2-Bromophenyl isothiocyanate

Purity: 98% CAS No.: 13037-60-0 Molecular Formula: C7H4BrNS Formula Weight: 214.09 Flash Point: 110°(230°F) Refractive Index: 1.6843 Odor: Pungent Boiling Point: 256-257° Beilstein Reference: 2802536 UN No.: 2922 MDL No.: MFCD00004799

Formula: C₇H₄BrNS
Boiling Pt: 256…257 °C
Melting Pt: 20 °C
Density: 1.591
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004799
CAS Number: 13037-60-0
UN: 2922
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR