Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L09968-14
L09968-22
L09968-36
CAAAAL09968-14EA
48.19
CAD
CAAAAL09968-14
CAAAAL09968-22
CAAAAL09968-36
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one 98%
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one
Purity: 98%
CAS No.: 7226-23-5
Molecular Formula: C6H12N2O
Formula Weight: 128.18
Flash Point: 120°(248°F)
Refractive Index: 1.4890
Odor: Odorless
pH: 11
Melting Point: -23°
Boiling Point: 246-247°
Beilstein Reference: 110562
MDL No.: MFCD00006550
Formula:
C₆H₁₂N₂O Boiling Pt: 246…247 °C Melting Pt: –23 °C Density: 1.060 Flash Pt: 120 °C (248 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006550 CAS Number: 7226-23-5 EINECS: 230-625-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...