4-Butylphenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09957-03
L09957-14
CAAAAL09957-03EA
50.4
CAD
CAAAAL09957-03
CAAAAL09957-14
4-Butylphenyl isocyanate 98%
4-Butylphenyl isocyanate
Purity: 98%
CAS No.: 69342-47-8
Molecular Formula: C11H13NO
Formula Weight: 175.23
Storage Temperature: 2°C to 8°C
Flash Point: 108°(226°F)
Refractive Index: 1.5165
Boiling Point: 107-108°/3mm
Beilstein Reference: 390859
UN No.: 2206
MDL No.: MFCD00013882
Formula:
C₁₁H₁₃NO Boiling Pt: 107…108 °C/3 mm Density: 0.992 Flash Pt: 108 °C (226 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013882 CAS Number: 69342-47-8 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...