n-Hexane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexyl hydride, Hexanen, Hexane
L09938-AD
L09938-AK
L09938-AU
CAAAAL09938-ADEA
60.51
CAD
CAAAAL09938-AD
CAAAAL09938-AK
CAAAAL09938-AU
n-Hexane 99%
n-Hexane
Purity: 99%
CAS No.: 110-54-3
Molecular Formula: CH3(CH2)4CH3
Formula Weight: 86.18
Flash Point: -23°(-9°F)
Refractive Index: 1.3750
Odor: Weak, characteristic
Melting Point: -95°
Boiling Point: 69°
Beilstein Reference: 1730733
Merck Reference: 14,4694
UN No.: 1208
MDL No.: MFCD00009520
Formula:
H₃C(CH₂)₄CH₃ MW: 86.18 g/mol Boiling Pt: 69 °C Melting Pt: –95 °C Density: 0.659 Flash Pt: –23 °C (–9 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009520 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 14,4694 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...