DL-Arabinose 98+%

Supplier: Thermo Scientific Chemicals

L09895-09 L09895-18
CAAAAL09895-09EA 151.21 CAD
CAAAAL09895-09 CAAAAL09895-18
DL-Arabinose 98+%
DL-Arabinose

Purity: 98+% CAS No.: 147-81-9 Molecular Formula: C5H10O5 Formula Weight: 150.13 Melting Point: 158-160° Beilstein Reference: 1723086 Merck Reference: 14,761 MDL No.: MFCD00135867

Formula: C₅H₁₀O₅
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 158…160 °C
Density: 1.609 g/cm³ (17 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135867
CAS Number: 147-81-9
EINECS: 205-699-8
Merck Index: 14,761

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR