3-Bromophenyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-3-isothiocyanatobenzene

L09871-06 L09871-14
CAAAAL09871-06EA 73.47 CAD
CAAAAL09871-06 CAAAAL09871-14
3-Bromophenyl isothiocyanate 97%
3-Bromophenyl isothiocyanate

Purity: 97% CAS No.: 2131-59-1 Molecular Formula: C7H4BrNS Formula Weight: 214.09 Flash Point: 110°(230°F) Refractive Index: 1.6850 Odor: Pungent Boiling Point: 255-256° Beilstein Reference: 2081173 UN No.: 2922 MDL No.: MFCD00004803

Formula: C₇H₄BrNS
MW: 214.09 g/mol
Boiling Pt: 255…256 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004803
CAS Number: 2131-59-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR