4-Chloro-2-(trifluoromethyl)phenyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

L09850-03 L09850-06
CAAAAL09850-03EA 56.88 CAD
CAAAAL09850-03 CAAAAL09850-06
4-Chloro-2-(trifluoromethyl)phenyl isocyanate 97%
4-Chloro-2-(trifluoromethyl)phenyl isocyanate

Purity: 97% CAS No.: 16588-69-5 Molecular Formula: C8H3ClF3NO Formula Weight: 221.57 Storage Temperature: 2°C to 8°C Flash Point: 94°(201°F) Refractive Index: 1.5010 Odor: Pungent Boiling Point: 114-115°/10mm Beilstein Reference: 2214336 UN No.: 2206 MDL No.: MFCD00013873

Formula: C₈H₃ClF₃NO
Boiling Pt: 114…115 °C/10 mm
Density: 1.467
Flash Pt: 94 °C (201 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00013873
CAS Number: 16588-69-5
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR