1,4-Phenylene diisothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Diisothiocyanatobenzene, p-Diisothiocyanatobenzene, Phenyl 1,4-diisothiocyanate, Bitoscanate, p-Phenylene diisothiocyanate, PDITC

L09756-03 L09756-06 L09756-14
CAAAAL09756-03EA 63.04 CAD
CAAAAL09756-03 CAAAAL09756-06 CAAAAL09756-14
1,4-Phenylene diisothiocyanate 98%
1,4-Phenylene diisothiocyanate

Purity: 98% CAS No.: 4044-65-9 Molecular Formula: C8H4N2S2 Formula Weight: 192.26 Storage Temperature: 2°C to 8°C Odor: Irritating Beilstein Reference: 2414340 Merck Reference: 14,1308 UN No.: 2923 MDL No.: MFCD00004811

Formula: C₈H₄N₂S₂
MW: 192.27 g/mol
Boiling Pt: 319
Melting Pt: 131
Storage Temperature: Refrigerator
MDL Number: MFCD00004811
CAS Number: 4044-65-9
EINECS: 223-741-3
UN: 2811
ADR: 6.1 III
Merck Index: 14,1308

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR