3,5-Dimethylbenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09748-03
L09748-06
CAAAAL09748-03EA
82.48
CAD
CAAAAL09748-03
CAAAAL09748-06
CAAAAL09748-14
3,5-Dimethylbenzaldehyde 98%
3,5-Dimethylbenzaldehyde
Purity: 98+%
CAS No.: 5779-95-3
Molecular Formula: C9H10O
Formula Weight: 134.18
Refractive Index: 1.5375
Melting Point: 8-10°
Boiling Point: 220-222°
Beilstein Reference: 2038840
MDL No.: MFCD00082777
Formula:
C₉H₁₀O Boiling Pt: 220…222 °C Melting Pt: 8…10 °C Density: 1.09 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00082777 CAS Number: 5779-95-3 EINECS: 675-424-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...