Tetramethoxysilane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L09716-14
L09716-22
L09716-36
CAAAAL09716-14EA
50.09
CAD
CAAAAL09716-14
CAAAAL09716-22
CAAAL09716-36
Tetramethoxysilane 98%
Tetramethoxysilane
Purity: 98%
CAS No.: 681-84-5
Molecular Formula: C4H12O4Si
Formula Weight: 152.22
Flash Point: 26°(78°F)
Refractive Index: 1.3680
Odor: Ether-like
Melting Point: 4-5°
Boiling Point: 121-122°
Beilstein Reference: 1699658
UN No.: 2606
MDL No.: MFCD00008341
Formula:
C₄H₁₂O₄Si Boiling Pt: 121…122 °C Melting Pt: 4…5 °C Density: 1.032 Flash Pt: 26 °C(78 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008341 CAS Number: 681-84-5 EINECS: 211-656-4 UN: 2606 ADR: "6.1",I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...