3,5-Xylylacetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09707-03
L09707-06
CAAAAL09707-03EA
89.43
CAD
CAAAAL09707-03
CAAAAL09707-06
3,5-Xylylacetic acid 98+%
3,5-Xylylacetic acid
Purity: 98+%
CAS No.: 42288-46-0
Molecular Formula: C10H12O2
Formula Weight: 164.21
Melting Point: 99-103°
Beilstein Reference: 2083903
Formula:
(C₆H₃(CH₃)₂)CH₂COOH Melting Pt: 99…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082776 CAS Number: 42288-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...