2-Methoxythiophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09689-06
L09689-14
CAAAAL09689-06EA
89.59
CAD
CAAAAL09689-06
CAAAAL09689-14
2-Methoxythiophenol 97%
2-Methoxythiophenol
Purity: 97%
CAS No.: 7217-59-6
Molecular Formula: C7H8OS
Formula Weight: 140.21
Flash Point: 85°(185°F)
Refractive Index: 1.5918
Odor: Unpleasant
Boiling Point: 225-228°
Beilstein Reference: 2042178
MDL No.: MFCD00004834
Formula:
C₇H₈OS Boiling Pt: 225…228 °C Density: 1.152 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004834 CAS Number: 7217-59-6 EINECS: 230-605-7 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...