2-Amino-4-(trifluoromethyl)thiophenol hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Amino-4-(trifluoromethyl)benzenethiol hydrochloride

L09683-03 L09683-06
CAAAAL09683-03EA 48.03 CAD
CAAAAL09683-03 CAAAAL09683-06
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride 97%
2-Amino-4-(trifluoromethyl)thiophenol hydrochloride

Purity: 97% CAS No.: 4274-38-8 Molecular Formula: C7H6F3NS.HCl Formula Weight: 229.65 Melting Point: 186-190° Beilstein Reference: 4273774 UN No.: 1759 MDL No.: MFCD00042600

Formula: C₇H₇ClF₃NS
Melting Pt: 186…190 °C
Density: 2.46
Storage Temperature: Ambient
MDL Number: MFCD00042600
CAS Number: 4274-38-8
EINECS: 429-560-8
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR