Tetramethylammonium hydroxide pentahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

L09658-14 L09658-22
CAAAAL09658-14EA 95.12 CAD
CAAAAL09658-14 CAAAAL09658-22
Tetramethylammonium hydroxide pentahydrate 98%
Tetramethylammonium hydroxide pentahydrate

Purity: 98% CAS No.: 10424-65-4 Molecular Formula: C4H13NO.5H2O Formula Weight: 181.23 (91.15anhy) Melting Point: 66-67° Beilstein Reference: 3714235 Merck Reference: 14,9224 UN No.: 3423 MDL No.: MFCD00149566

Formula: C₄H₁₃NO·5H₂O
Melting Pt: 66…67 °C
Density: 1.829 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149566
CAS Number: 10424-65-4
EINECS: 200-882-9
UN: 3423
ADR: 8,II
Merck Index: 14,9224

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR