3-(Trimethylsilyl)-2-propyn-1-ol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(Trimethylsilyl)propargyl alcohol
L09649-03
L09649-06
L09649-14
CAAAAL09649-03EA
62.73
CAD
CAAAAL09649-03
CAAAAL09649-06
CAAAAL09649-14
3-(Trimethylsilyl)-2-propyn-1-ol 98+%
3-(Trimethylsilyl)-2-propyn-1-ol
Purity: 98+%
CAS No.: 5272-36-6
Molecular Formula: C6H12OSi
Formula Weight: 128.25
Storage Temperature: 2°C to 8°C
Flash Point: 66°(150°F)
Refractive Index: 1.4510
Boiling Point: 170-172°
Beilstein Reference: 1902505
UN No.: 2810
MDL No.: MFCD00002913
Formula:
C₆H₁₂OSi MW: 128.25 g/mol Boiling Pt: 170…172 °C Density: 0.865 Flash Pt: 66 °C (150 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002913 CAS Number: 5272-36-6 EINECS: 226-094-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...