Guanidinium nitrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Guanidine nitrate

L09643-0B L09643-30
CAAAAL09643-0BEA 147.1 CAD
CAAAAL09643-0B CAAAAL09643-30
Guanidinium nitrate 98%
Guanidinium nitrate

Purity: 98% CAS No.: 506-93-4 Molecular Formula: CH6N4O3 Formula Weight: 122.08 Odor: Odorless pH: 4.9 Melting Point: 213-216° Beilstein Reference: 3596600 Merck Reference: 14,4562 UN No.: 1467 MDL No.: MFCD00013028

Formula: CH₆N₄O₃
MW: 122.08 g/mol
Melting Pt: 213…216 °C
Density: 1.44
Storage Temperature: Ambient
MDL Number: MFCD00013028
CAS Number: 506-93-4
EINECS: 208-060-1
UN: 1467
ADR: 5.1,III
Merck Index: 14,4562

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR