Methyl-3,4-dihydroxybenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09552-04
L09552-09
CAAAAL09552-04EA
80.26
CAD
CAAAAL09552-04
CAAAAL09552-09
Methyl-3,4-dihydroxybenzoate 97%
Methyl 3,4-dihydroxybenzoate
Purity: 97%
CAS No.: 2150-43-8
Molecular Formula: C8H8O4
Formula Weight: 168.15
Odor: Odorless
Melting Point: 134-138°
Beilstein Reference: 1637939
MDL No.: MFCD00016437
Formula:
C₈H₈O₄ Melting Pt: 134…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016437 CAS Number: 2150-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...