(±)-1-Phenyl-2-propyn-1-ol 98%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09549-03
L09549-06
CAAAAL09549-03EA
49.61
CAD
CAAAAL09549-03
CAAAAL09549-06
(±)-1-Phenyl-2-propyn-1-ol 98%, Technical Grade
(±)-1-Phenyl-2-propyn-1-ol
Purity: 98%
CAS No.: 4187-87-5
Molecular Formula: C9H8O
Formula Weight: 132.16
Flash Point: 99°(210°F)
Refractive Index: 1.5500
Melting Point: 26-30°
Boiling Point: 231-233°
Beilstein Reference: 742365
MDL No.: MFCD00021860
Formula:
C₉H₈O Boiling Pt: 231…233 °C Melting Pt: 26…30 °C Density: 1.087 Flash Pt: 99 °C(210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00021860 CAS Number: 4187-87-5 EINECS: 224-064-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...