2-Butoxyethyl acetate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09515-0F
L09515-AE
L09515-AP
CAAAAL09515-0FEA
195.92
CAD
CAAAAL09515-0F
CAAAAL09515-AE
CAAAAL09515-AP
2-Butoxyethyl acetate 98%
2-Butoxyethyl acetate
Purity: 98%
CAS No.: 112-07-2
Molecular Formula: C8H16O3
Formula Weight: 160.21
Flash Point: 71°(159°F)
Refractive Index: 1.4130
Odor: Fruit-like
Melting Point: -64°
Boiling Point: 191-192°
Beilstein Reference: 1756960
MDL No.: MFCD00009457
Formula:
C₈H₁₆O₃ Boiling Pt: 191…192 °C Melting Pt: –64 °C Density: 0,942 Flash Pt: 71 °C(159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009457 CAS Number: 112-07-2 EINECS: 203-933-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...