2,3-Dichlorobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

L09506-06 L09506-14 L09506-22
CAAAAL09506-06EA 77.74 CAD
CAAAAL09506-06 CAAAAL09506-14 CAAAAL09506-22
2,3-Dichlorobenzoyl chloride 98%
2,3-Dichlorobenzoyl chloride

Purity: 98% CAS No.: 2905-60-4 Molecular Formula: C7H3Cl3O Formula Weight: 209.46 Flash Point: 167°(332°F) Odor: Pungent Boiling Point: 140°/14mm Beilstein Reference: 2575973 UN No.: 3261 MDL No.: MFCD00035937

Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 140 °C/14 mm
Melting Pt: 28…32 °C
Flash Pt: 167 °C (332 °F)
Storage Temperature: Ambient
MDL Number: MFCD00035937
CAS Number: 2905-60-4
EINECS: 220-811-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR