4-Amino-3-nitrophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09487-06
L09487-14
CAAAAL09487-06EA
52.61
CAD
CAAAAL09487-06
CAAAAL09487-14
CAAAAL09487-22
4-Amino-3-nitrophenol 98%
4-Amino-3-nitrophenol
Purity: 98%
CAS No.: 610-81-1
Molecular Formula: C6H6N2O3
Formula Weight: 154.13
Odor: Characteristic
Melting Point: 148-153°
Beilstein Reference: 2210196
MDL No.: MFCD00066310
Formula:
C₆H₆N₂O₃ Melting Pt: 148…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066310 CAS Number: 610-81-1 EINECS: 210-236-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...