(2-Bromoethyl)cyclohexane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09466-03
L09466-06
L09466-14
CAAAAL09466-03EA
43.45
CAD
CAAAAL09466-03
CAAAAL09466-06
CAAAAL09466-14
(2-Bromoethyl)cyclohexane 99%
(2-Bromoethyl)cyclohexane
Purity: 99%
CAS No.: 1647-26-3
Molecular Formula: C8H15Br
Formula Weight: 191.12
Flash Point: 96°(204°F)
Refractive Index: 1.4900
Boiling Point: 70-71°/6mm
Beilstein Reference: 1919816
MDL No.: MFCD00019398
Formula:
C₈H₁₅Br Boiling Pt: 70…71 °C (6 mmHg) Density: 1,221 Flash Pt: 96 °C (204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019398 CAS Number: 1647-26-3 EINECS: 216-712-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...