3-Cyanophenyl isocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L09432-03
L09432-06
CAAAAL09432-03EA
50.88
CAD
CAAAAL09432-03
CAAAAL09432-06
3-Cyanophenyl isocyanate 97%
3-Cyanophenyl isocyanate
Purity: 97%
CAS No.: 16413-26-6
Molecular Formula: C8H4N2O
Formula Weight: 144.13
Storage Temperature: 2°C to 8°C
Odor: Irritating
Beilstein Reference: 3089117
UN No.: 2811
MDL No.: MFCD00013856
Formula:
C₈H₄N₂O Boiling Pt: 123 °C (12 -rr) Melting Pt: 51…54 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013856 CAS Number: 16413-26-6 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...